Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003054.4 | NP_003045.2 | 7 - 8 | 951 | 55 |
GenBank mRNA | AK314997.1 | - | 7 - 8 | 904 | 55 |
BC030593.1 | - | 6 - 7 | 1209 | 55 | |
BC108927.1 | - | 6 - 7 | 816 | 55 | |
BC108928.2 | - | 6 - 7 | 816 | 55 | |
L09118.1 | - | 7 - 8 | 893 | 55 | |
L14269.1 | - | 7 - 8 | 821 | 55 | |
L23205.1 | - | 7 - 8 | 901 | 55 | |
X71354.1 | - | 7 - 8 | 851 | 55 |