Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322278.1 | NP_001309207.1 | 9 - 10 | 1090 | 85 |
NM_001322279.1 | NP_001309208.1 | 9 - 10 | 1054 | 85 | |
NM_199227.2 | NP_954697.1 | 9 - 10 | 1004 | 85 | |
NR_136273.1 | 8 - 9 | 941 | 85 | ||
NR_136276.1 | 8 - 9 | 855 | 85 | ||
GenBank mRNA | AY374142.1 | - | 9 - 10 | 1004 | 85 |
BC113644.1 | - | 9 - 10 | 949 | 85 | |
BC143913.1 | - | 9 - 10 | 1035 | 85 | |
DQ005576.1 | - | 9 - 10 | 1318 | 85 |