Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005765.2 | NP_005756.2 | 3 - 4 | 407 | 94 |
GenBank mRNA | AF109363.1 | - | 3 - 4 | 368 | 94 |
AF248966.1 | - | 3 - 4 | 412 | 94 | |
AF291814.1 | - | 3 - 4 | 394 | 94 | |
AK075382.1 | - | 3 - 4 | 465 | 94 | |
AK296614.1 | - | 2 - 3 | 270 | 94 | |
AK297007.1 | - | 3 - 4 | 378 | 94 | |
AK314633.1 | - | 3 - 4 | 358 | 94 | |
AL049929.1 | - | 2 - 3 | 247 | 94 | |
AY038990.1 | - | 3 - 4 | 410 | 94 | |
AY429341.1 | - | 2 - 3 | 270 | 94 | |
BC084541.1 | - | 3 - 4 | 387 | 94 |