Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005765.2 | NP_005756.2 | 5 - 6 | 642 | 111 |
GenBank mRNA | AF109363.1 | - | 5 - 6 | 604 | 111 |
AF248966.1 | - | 5 - 6 | 647 | 111 | |
AF291814.1 | - | 5 - 6 | 629 | 111 | |
AK075382.1 | - | 5 - 6 | 700 | 111 | |
AK296614.1 | - | 4 - 5 | 505 | 111 | |
AK297760.1 | - | 4 - 5 | 616 | 111 | |
AK314633.1 | - | 5 - 6 | 593 | 111 | |
AK315948.1 | - | 4 - 5 | 477 | 111 | |
AL049929.1 | - | 4 - 5 | 482 | 111 | |
AY038990.1 | - | 5 - 6 | 645 | 111 | |
AY429341.1 | - | 4 - 5 | 505 | 111 | |
BC010395.1 | - | 1 - 2 | 102 | 111 | |
BC084541.1 | - | 5 - 6 | 622 | 111 |