Assay Design



Gene Transcripts

Gene Symbol
BZW2
Entrez Gene ID
28969
Gene Name
basic leucine zipper and W2 domains 2
Gene Aliases
HSPC028, MST017, MSTP017
Chromosome Location
Chr.7: 16646134 - 16706523 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159767.1 NP_001153239.1 7 - 8 827 93
NM_014038.2 NP_054757.1 7 - 8 811 93
NR_027624.1 6 - 7 691 93
XM_006715706.1 XP_006715769.1 7 - 8 695 93
XM_006715707.1 XP_006715770.1 7 - 8 778 93
XM_006715708.1 XP_006715771.1 7 - 8 697 93
GenBank mRNA AF083246.1 - 7 - 8 811 93
AK001218.1 - 7 - 8 791 93
AK001260.1 - 7 - 8 785 93
AK297496.1 - 7 - 8 740 93
BC001570.1 - 3 - 4 291 93
BC003056.1 - 7 - 8 737 93
BC008453.1 - 7 - 8 764 93
BC009597.1 - 7 - 8 795 93
BC017794.1 - 7 - 8 663 93
BC107757.1 - 7 - 8 785 93

View Full Product Details