Assay Design
Gene Transcripts
- Gene Symbol
- C2orf54
- Entrez Gene ID
-
79919
- Gene Name
- chromosome 2 open reading frame 54
- Gene Aliases
-
-
- Chromosome Location
-
Chr.2: 240886048 - 240896889 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_024861.3 |
NP_079137.3 |
1 - 2 |
105 |
131 |
View Full Product Details