Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000489.4 | NP_000480.3 | 24 - 25 | 6045 | 106 |
NM_138270.3 | NP_612114.2 | 23 - 24 | 5931 | 106 | |
XM_005262153.4 | XP_005262210.2 | 24 - 25 | 6042 | 106 | |
XM_005262154.4 | XP_005262211.2 | 25 - 26 | 5958 | 106 | |
XM_005262155.4 | XP_005262212.2 | 24 - 25 | 6146 | 106 | |
XM_005262156.4 | XP_005262213.2 | 23 - 24 | 5898 | 106 | |
XM_005262157.4 | XP_005262214.2 | 24 - 25 | 5841 | 106 | |
XM_006724666.3 | XP_006724729.1 | 23 - 24 | 5928 | 106 | |
XM_006724667.3 | XP_006724730.1 | 22 - 23 | 5784 | 106 | |
XM_017029601.1 | XP_016885090.1 | 25 - 26 | 5955 | 106 | |
XM_017029602.1 | XP_016885091.1 | 24 - 25 | 6126 | 106 | |
XM_017029603.1 | XP_016885092.1 | 23 - 24 | 5895 | 106 | |
XM_017029604.1 | XP_016885093.1 | 24 - 25 | 5844 | 106 | |
XM_017029605.1 | XP_016885094.1 | 25 - 26 | 6059 | 106 | |
XM_017029606.1 | XP_016885095.1 | 23 - 24 | 6032 | 106 | |
XM_017029607.1 | XP_016885096.1 | 23 - 24 | 6029 | 106 | |
XM_017029608.1 | XP_016885097.1 | 22 - 23 | 5781 | 106 | |
XM_017029609.1 | XP_016885098.1 | 24 - 25 | 5945 | 106 | |
XM_017029610.1 | XP_016885099.1 | 24 - 25 | 5942 | 106 | |
XM_017029611.1 | XP_016885100.1 | 23 - 24 | 5697 | 106 | |
GenBank mRNA | AB102641.1 | - | 24 - 25 | 5777 | 106 |
AK295107.1 | - | 7 - 8 | 897 | 106 | |
AK296959.1 | - | 3 - 4 | 280 | 106 | |
U09820.1 | - | 16 - 17 | 3256 | 106 | |
U72936.2 | - | 25 - 26 | 6114 | 106 | |
U72937.2 | - | 24 - 25 | 5992 | 106 | |
U72938.2 | - | 23 - 24 | 5878 | 106 |