Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000489.4 | NP_000480.3 | 34 - 35 | 7471 | 109 |
NM_138270.3 | NP_612114.2 | 33 - 34 | 7357 | 109 | |
XM_005262153.4 | XP_005262210.2 | 34 - 35 | 7468 | 109 | |
XM_005262154.4 | XP_005262211.2 | 35 - 36 | 7384 | 109 | |
XM_005262155.4 | XP_005262212.2 | 34 - 35 | 7572 | 109 | |
XM_005262156.4 | XP_005262213.2 | 33 - 34 | 7324 | 109 | |
XM_005262157.4 | XP_005262214.2 | 34 - 35 | 7267 | 109 | |
XM_006724666.3 | XP_006724729.1 | 33 - 34 | 7354 | 109 | |
XM_006724667.3 | XP_006724730.1 | 32 - 33 | 7210 | 109 | |
XM_017029601.1 | XP_016885090.1 | 35 - 36 | 7381 | 109 | |
XM_017029602.1 | XP_016885091.1 | 34 - 35 | 7552 | 109 | |
XM_017029603.1 | XP_016885092.1 | 33 - 34 | 7321 | 109 | |
XM_017029604.1 | XP_016885093.1 | 34 - 35 | 7270 | 109 | |
XM_017029605.1 | XP_016885094.1 | 35 - 36 | 7485 | 109 | |
XM_017029606.1 | XP_016885095.1 | 33 - 34 | 7458 | 109 | |
XM_017029607.1 | XP_016885096.1 | 33 - 34 | 7455 | 109 | |
XM_017029608.1 | XP_016885097.1 | 32 - 33 | 7207 | 109 | |
XM_017029609.1 | XP_016885098.1 | 34 - 35 | 7371 | 109 | |
XM_017029610.1 | XP_016885099.1 | 34 - 35 | 7368 | 109 | |
XM_017029611.1 | XP_016885100.1 | 33 - 34 | 7123 | 109 | |
GenBank mRNA | AB102641.1 | - | 34 - 35 | 7203 | 109 |
AK295107.1 | - | 17 - 18 | 2323 | 109 | |
AK296959.1 | - | 13 - 14 | 1706 | 109 | |
AK297183.1 | - | 6 - 7 | 780 | 109 | |
BX647222.1 | - | 7 - 8 | 5525 | 109 | |
U72936.2 | - | 35 - 36 | 7540 | 109 | |
U72937.2 | - | 34 - 35 | 7418 | 109 | |
U72938.2 | - | 33 - 34 | 7304 | 109 |