Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014467.2 | NP_055282.1 | 4 - 5 | 834 | 76 |
GenBank mRNA | AF060567.1 | - | 4 - 5 | 772 | 76 |
AF393649.1 | - | 4 - 5 | 693 | 76 | |
AK075462.1 | - | 4 - 5 | 782 | 76 | |
BC020733.1 | - | 4 - 5 | 609 | 76 |