Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166222.1 | NP_001159694.1 | 6 - 7 | 1140 | 96 |
NM_020811.1 | NP_065862.1 | 5 - 6 | 1113 | 96 | |
XM_011545189.2 | XP_011543491.1 | 5 - 6 | 1430 | 96 | |
XM_011545191.2 | XP_011543493.1 | 4 - 5 | 2642 | 96 | |
XM_017018054.1 | XP_016873543.1 | 5 - 6 | 2344 | 96 | |
XM_017018055.1 | XP_016873544.1 | 3 - 4 | 1624 | 96 | |
GenBank mRNA | AB037815.1 | - | 4 - 5 | 2238 | 96 |
AK294035.1 | - | 6 - 7 | 1140 | 96 | |
EF560746.1 | - | 5 - 6 | 1118 | 96 |