Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014067.3 | NP_054786.2 | 10 - 11 | 1128 | 135 |
GenBank mRNA | AF202922.2 | - | 10 - 11 | 1061 | 135 |
BC000270.1 | - | 10 - 11 | 1076 | 135 | |
BC003188.1 | - | 10 - 11 | 882 | 135 | |
BC007297.1 | - | 10 - 11 | 1076 | 135 | |
BC008316.1 | - | 10 - 11 | 1076 | 135 |