Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000021.3 | NP_000012.1 | 5 - 6 | 767 | 141 |
NM_007318.2 | NP_015557.2 | 5 - 6 | 755 | 141 | |
XM_005267864.2 | XP_005267921.1 | 5 - 6 | 761 | 141 | |
XM_005267866.1 | XP_005267923.1 | 5 - 6 | 750 | 141 | |
XM_011536971.2 | XP_011535273.1 | 5 - 6 | 659 | 141 | |
XM_011536972.2 | XP_011535274.1 | 5 - 6 | 1023 | 141 | |
XM_011536973.1 | XP_011535275.1 | 5 - 6 | 1011 | 141 | |
XM_011536974.1 | XP_011535276.1 | 5 - 6 | 652 | 141 | |
GenBank mRNA | AF416717.1 | - | 3 - 4 | 483 | 141 |
AK122722.1 | - | 3 - 4 | 2036 | 141 | |
AK223173.1 | - | 3 - 4 | 497 | 141 | |
AK312531.1 | - | 5 - 6 | 751 | 141 | |
BC011729.2 | - | 5 - 6 | 736 | 141 | |
KU178285.1 | - | 3 - 4 | 471 | 141 | |
KU178286.1 | - | 3 - 4 | 483 | 141 | |
L42110.1 | - | 5 - 6 | 731 | 141 | |
L76517.1 | - | 5 - 6 | 1024 | 141 | |
U40379.1 | - | 3 - 4 | 471 | 141 | |
U40380.1 | - | 3 - 4 | 471 | 141 |