Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172745.1 | NP_001166216.1 | 16 - 17 | 2254 | 73 |
NM_001172746.1 | NP_001166217.1 | 16 - 17 | 2290 | 73 | |
NM_006363.4 | NP_006354.2 | 16 - 17 | 2348 | 73 | |
NM_032985.4 | NP_116780.1 | 16 - 17 | 2366 | 73 | |
NM_032986.3 | NP_116781.1 | 16 - 17 | 2045 | 73 | |
XM_017027593.1 | XP_016883082.1 | 16 - 17 | 2242 | 73 | |
GenBank mRNA | AK296270.1 | - | 16 - 17 | 1966 | 73 |
AK299513.1 | - | 12 - 13 | 1246 | 73 | |
BC001575.1 | - | 16 - 17 | 1961 | 73 | |
BC005032.1 | - | 16 - 17 | 2008 | 73 | |
BC005404.1 | - | 16 - 17 | 2155 | 73 | |
BC095404.1 | - | 16 - 17 | 2002 | 73 | |
X97065.1 | - | 16 - 17 | 2010 | 73 |