Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172745.1 | NP_001166216.1 | 4 - 5 | 711 | 89 |
NM_006363.4 | NP_006354.2 | 4 - 5 | 805 | 89 | |
NM_032985.4 | NP_116780.1 | 4 - 5 | 823 | 89 | |
NM_032986.3 | NP_116781.1 | 4 - 5 | 502 | 89 | |
XM_017027593.1 | XP_016883082.1 | 4 - 5 | 699 | 89 | |
GenBank mRNA | AK299171.1 | - | 4 - 5 | 424 | 89 |
AK299513.1 | - | 4 - 5 | 427 | 89 | |
AL540773.3 | - | 4 - 5 | 482 | 89 | |
BC001575.1 | - | 4 - 5 | 419 | 89 | |
BC005032.1 | - | 4 - 5 | 465 | 89 | |
BC005404.1 | - | 4 - 5 | 612 | 89 | |
BC095404.1 | - | 4 - 5 | 459 | 89 | |
BM751928.1 | - | 4 - 5 | 502 | 89 | |
BX398403.2 | - | 4 - 5 | 826 | 89 | |
X97065.1 | - | 4 - 5 | 467 | 89 |