Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030755.4 | NP_110382.3 | 1 - 2 | 273 | 102 |
GenBank mRNA | AB048246.1 | - | 1 - 2 | 265 | 102 |
AK075395.1 | - | 1 - 2 | 180 | 102 | |
AK302416.1 | - | 1 - 2 | 225 | 102 | |
AK303134.1 | - | 1 - 2 | 167 | 102 | |
AK312905.1 | - | 1 - 2 | 151 | 102 | |
AL080080.1 | - | 1 - 2 | 171 | 102 | |
AY358640.1 | - | 1 - 2 | 160 | 102 | |
BC036460.2 | - | 1 - 2 | 223 | 102 | |
BC056874.1 | - | 1 - 2 | 162 | 102 | |
DA203669.1 | - | 1 - 2 | 273 | 102 |