Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030755.4 | NP_110382.3 | 6 - 7 | 713 | 88 |
GenBank mRNA | AB048246.1 | - | 6 - 7 | 705 | 88 |
AK075395.1 | - | 6 - 7 | 620 | 88 | |
AK303134.1 | - | 6 - 7 | 703 | 88 | |
AK312905.1 | - | 6 - 7 | 591 | 88 | |
AL080080.1 | - | 6 - 7 | 611 | 88 | |
AY358640.1 | - | 6 - 7 | 600 | 88 | |
BC036460.2 | - | 6 - 7 | 663 | 88 | |
BC056874.1 | - | 6 - 7 | 602 | 88 |