Assay Design



Gene Transcripts

Gene Symbol
MAP4K5
Entrez Gene ID
11183
Gene Name
mitogen-activated protein kinase kinase kinase kinase 5
Gene Aliases
GCKR, KHS, KHS1, MAPKKKK5
Chromosome Location
Chr.14: 50418493 - 50561126 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006575.4 NP_006566.2 1 - 2 207 103
XM_011536377.2 XP_011534679.1 1 - 2 116 103
GenBank mRNA CB959746.1 - - 103
CN343360.1 - 1 - 2 140 103
U77129.1 - 1 - 2 207 103

View Full Product Details