Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052848.1 | NP_443080.1 | 2 - 3 | 626 | 71 |
XM_005259377.4 | XP_005259434.1 | 2 - 3 | 989 | 71 | |
XM_017027442.1 | XP_016882931.1 | 2 - 3 | 960 | 71 | |
GenBank mRNA | AL833726.1 | - | 1 - 2 | 461 | 71 |
BC011577.1 | - | 2 - 3 | 626 | 71 |