Assay Design



Gene Transcripts

Gene Symbol
CCDC97
Entrez Gene ID
90324
Gene Name
coiled-coil domain containing 97
Gene Aliases
-
Chromosome Location
Chr.19: 41309855 - 41324883 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_052848.1 NP_443080.1 4 - 5 1026 91
XM_005259377.4 XP_005259434.1 4 - 5 1389 91
GenBank mRNA AK097586.1 - 2 - 3 189 91
AL833726.1 - 3 - 4 861 91
BC007575.1 - 2 - 3 396 91
BC011577.1 - 4 - 5 1026 91

View Full Product Details