Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190821.1 | NP_001177750.1 | 1 - 2 | 895 | 122 |
NM_005904.3 | NP_005895.1 | 1 - 2 | 895 | 122 | |
GenBank mRNA | AF010193.1 | - | 1 - 2 | 903 | 122 |
AF015261.1 | - | 1 - 2 | 608 | 122 | |
BC074818.2 | - | 1 - 2 | 642 | 122 | |
BC074819.2 | - | 1 - 2 | 642 | 122 | |
DA882147.1 | - | 1 - 2 | 327 | 122 |