Assay Design



Gene Transcripts

Gene Symbol
GID8
Entrez Gene ID
54994
Gene Name
GID complex subunit 8 homolog
Gene Aliases
C20orf11, TWA1
Chromosome Location
Chr.20: 62938089 - 62948475 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017896.2 NP_060366.1 3 - 4 493 68
GenBank mRNA AK000609.1 - 3 - 4 429 68
BC032120.2 - 3 - 4 463 68

View Full Product Details