Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000973.4 | NP_000964.1 | 5 - 6 | 714 | 66 |
NM_001317771.1 | NP_001304700.1 | 5 - 6 | 757 | 66 | |
NM_001317782.1 | NP_001304711.1 | 4 - 5 | 882 | 66 | |
NM_033301.2 | NP_150644.1 | 5 - 6 | 778 | 66 | |
GenBank mRNA | AK289459.1 | - | 5 - 6 | 665 | 66 |
BC000047.2 | - | 3 - 4 | 486 | 66 | |
BC000077.2 | - | 5 - 6 | 649 | 66 | |
BC012197.1 | - | 5 - 6 | 733 | 66 | |
BC013104.1 | - | 4 - 5 | 646 | 66 | |
BC030151.2 | - | 4 - 5 | 636 | 66 | |
BC093064.1 | - | 5 - 6 | 688 | 66 | |
BI461661.1 | - | 5 - 6 | 718 | 67 | |
BM783910.1 | - | 2 - 3 | 290 | 66 | |
BT007379.1 | - | 4 - 5 | 622 | 66 | |
CR457046.1 | - | 4 - 5 | 622 | 66 | |
Z28407.1 | - | 5 - 6 | 665 | 66 |