Assay Design



Gene Transcripts

Gene Symbol
DICER1
Entrez Gene ID
23405
Gene Name
dicer 1, ribonuclease III
Gene Aliases
DCR1, Dicer, Dicer1e, HERNA, K12H4.8-LIKE, MNG1, RMSE2
Chromosome Location
Chr.14: 95086228 - 95158010 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030621.4 NP_085124.2 3 - 4 247 90
XM_011536599.1 XP_011534901.1 3 - 4 379 90
XM_011536600.2 XP_011534902.1 3 - 4 254 90
XM_011536602.2 XP_011534904.1 4 - 5 290 90
XM_017021120.1 XP_016876609.1 3 - 4 686 90
XM_017021122.1 XP_016876611.1 3 - 4 256 90
XM_017021123.1 XP_016876612.1 3 - 4 687 90
GenBank mRNA AB028449.1 - 2 - 3 137 90
AY929254.1 - 3 - 4 208 90
AY929256.1 - 4 - 5 242 90
BM152875.1 - 3 - 4 228 90

View Full Product Details