Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195573.1 | NP_001182502.1 | 3 - 4 | 481 | 73 |
NM_001271282.2 | NP_001258211.1 | 4 - 5 | 669 | 73 | |
NM_001291628.1 | NP_001278557.1 | 4 - 5 | 557 | 73 | |
NM_030621.4 | NP_085124.2 | 6 - 7 | 728 | 73 | |
NM_177438.2 | NP_803187.1 | 4 - 5 | 674 | 73 | |
XM_011536599.1 | XP_011534901.1 | 6 - 7 | 860 | 73 | |
XM_011536600.2 | XP_011534902.1 | 6 - 7 | 735 | 73 | |
XM_011536601.2 | XP_011534903.1 | 7 - 8 | 880 | 73 | |
XM_011536602.2 | XP_011534904.1 | 7 - 8 | 771 | 73 | |
XM_017021120.1 | XP_016876609.1 | 6 - 7 | 1167 | 73 | |
XM_017021121.1 | XP_016876610.1 | 4 - 5 | 562 | 73 | |
GenBank mRNA | AB023145.2 | - | 4 - 5 | 674 | 73 |
AB028449.1 | - | 5 - 6 | 624 | 73 | |
AJ132261.2 | - | 4 - 5 | 677 | 73 | |
BC150287.1 | - | 4 - 5 | 674 | 73 | |
BP199059.1 | - | 3 - 4 | 487 | 73 | |
HM595745.1 | - | 3 - 4 | 445 | 73 |