Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006035.3 | NP_006026.3 | 26 - 27 | 3686 | 71 |
XM_005268227.1 | XP_005268284.1 | 27 - 28 | 3567 | 71 | |
XM_005268228.1 | XP_005268285.1 | 26 - 27 | 3489 | 71 | |
XM_005268229.1 | XP_005268286.1 | 25 - 26 | 3438 | 71 | |
XM_005268230.4 | XP_005268287.1 | 27 - 28 | 3567 | 71 | |
XM_011537387.1 | XP_011535689.1 | 27 - 28 | 3567 | 71 | |
GenBank mRNA | AB032950.3 | - | 26 - 27 | 3677 | 71 |
AF128625.1 | - | 26 - 27 | 3743 | 71 | |
BC155541.1 | - | 26 - 27 | 3689 | 71 | |
BX248752.1 | - | 7 - 8 | 789 | 71 |