Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300895.1 | NP_001287824.1 | 4 - 5 | 799 | 86 |
NM_001300896.1 | NP_001287825.1 | 5 - 6 | 683 | 86 | |
NM_145200.3 | NP_660201.1 | 4 - 5 | 731 | 86 | |
XM_011545181.2 | XP_011543483.1 | 5 - 6 | 736 | 86 | |
XM_011545182.2 | XP_011543484.1 | 4 - 5 | 592 | 86 | |
XM_011545183.2 | XP_011543485.1 | 5 - 6 | 535 | 86 | |
XM_017018025.1 | XP_016873514.1 | 5 - 6 | 1021 | 86 | |
GenBank mRNA | AY039217.1 | - | 4 - 5 | 731 | 86 |
BC033167.1 | - | 5 - 6 | 683 | 86 |