Assay Design



Gene Transcripts

Gene Symbol
GAR1
Entrez Gene ID
54433
Gene Name
GAR1 ribonucleoprotein
Gene Aliases
NOLA1
Chromosome Location
Chr.4: 109815510 - 109824737 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018983.3 NP_061856.1 2 - 3 515 70
NM_032993.2 NP_127460.1 2 - 3 256 70
GenBank mRNA AJ276003.1 - 2 - 3 472 70
AK310489.1 - 2 - 3 398 70
AY780787.1 - 1 - 2 208 70
BC003413.1 - 2 - 3 256 70
CN428426.1 - 2 - 3 480 70

View Full Product Details