Assay Design



Gene Transcripts

Gene Symbol
GTDC1
Entrez Gene ID
79712
Gene Name
glycosyltransferase like domain containing 1
Gene Aliases
Hmat-Xa, mat-Xa
Chromosome Location
Chr.2: 143945787 - 144333092 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006636.3 NP_001006637.1 8 - 9 1264 85
NM_001164629.3 NP_001158101.1 8 - 9 1249 85
NM_001284234.1 NP_001271163.1 6 - 7 964 85
NM_001284238.1 NP_001271167.1 10 - 11 1463 85
XM_005263774.2 XP_005263831.1 9 - 10 1348 85
XM_005263782.2 XP_005263839.1 9 - 10 1364 85
XM_005263784.2 XP_005263841.1 7 - 8 1223 85
XM_011511843.2 XP_011510145.1 9 - 10 1397 85
XM_011511855.2 XP_011510157.1 7 - 8 1341 85
XM_011511856.2 XP_011510158.1 7 - 8 1123 85
XM_017004917.1 XP_016860406.1 13 - 14 2019 85
XM_017004918.1 XP_016860407.1 11 - 12 1611 85
XM_017004919.1 XP_016860408.1 9 - 10 1497 85
XM_017004920.1 XP_016860409.1 11 - 12 4653 85
XM_017004921.1 XP_016860410.1 10 - 11 1446 85
XM_017004922.1 XP_016860411.1 10 - 11 1623 85
XM_017004923.1 XP_016860412.1 9 - 10 1411 85
XM_017004924.1 XP_016860413.1 11 - 12 1607 85
XM_017004925.1 XP_016860414.1 8 - 9 1269 85
XM_017004926.1 XP_016860415.1 6 - 7 1200 85
XM_017004927.1 XP_016860416.1 8 - 9 1231 85
XM_017004928.1 XP_016860417.1 10 - 11 1464 85
XM_017004929.1 XP_016860418.1 8 - 9 1201 85
XM_017004930.1 XP_016860419.1 8 - 9 1852 85
XM_017004931.1 XP_016860420.1 8 - 9 1265 85
XM_017004932.1 XP_016860421.1 8 - 9 1235 85
XM_017004933.1 XP_016860422.1 8 - 9 1217 85
XM_017004934.1 XP_016860423.1 9 - 10 4424 85
XM_017004935.1 XP_016860424.1 10 - 11 2294 85
XM_017004936.1 XP_016860425.1 9 - 10 1471 85
XM_017004949.1 XP_016860438.1 5 - 6 845 85
XM_017004950.1 XP_016860439.1 7 - 8 892 85
XM_017004951.1 XP_016860440.1 7 - 8 847 85
XM_017004952.1 XP_016860441.1 7 - 8 1291 85
XM_017004953.1 XP_016860442.1 7 - 8 863 85
XM_017004954.1 XP_016860443.1 8 - 9 951 85
XM_017004955.1 XP_016860444.1 10 - 11 1299 85
GenBank mRNA AB201269.1 - 5 - 6 985 85
AB597184.1 - 5 - 6 985 85
AK021815.1 - 2 - 3 440 85
AK291357.1 - 8 - 9 1179 85
AY281366.1 - 8 - 9 1264 85
BX647436.1 - 6 - 7 946 85
BX647773.1 - 10 - 11 1402 85

View Full Product Details