Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020927.2 | NP_065978.1 | 4 - 5 | 845 | 82 |
GenBank mRNA | AB046796.1 | - | 4 - 5 | 815 | 82 |
AK290153.1 | - | 4 - 5 | 823 | 82 | |
AL834240.1 | - | 3 - 4 | 460 | 82 | |
BC033537.1 | - | 4 - 5 | 823 | 82 |