Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181453.3 | NP_852118.1 | 1 - 2 | 160 | 99 |
NR_028063.1 | 1 - 2 | 160 | 99 | ||
GenBank mRNA | AK090813.1 | - | 1 - 2 | 720 | 99 |
BC037774.2 | - | 1 - 2 | 101 | 99 | |
DA865539.1 | - | 1 - 2 | 162 | 101 |