Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284308.1 | NP_001271237.1 | 4 - 5 | 425 | 70 |
NM_001284309.1 | NP_001271238.1 | 4 - 5 | 365 | 70 | |
NM_001284310.1 | NP_001271239.1 | 3 - 4 | 340 | 70 | |
NM_001284311.1 | NP_001271240.1 | 2 - 3 | 193 | 70 | |
NM_006869.3 | NP_006860.1 | 4 - 5 | 598 | 70 | |
GenBank mRNA | AB097049.1 | - | 4 - 5 | 572 | 70 |
AF082324.1 | - | 4 - 5 | 572 | 70 | |
AJ006422.1 | - | 4 - 5 | 594 | 70 | |
AK092471.1 | - | 3 - 4 | 342 | 70 | |
AK290517.1 | - | 5 - 6 | 544 | 70 | |
AK300644.1 | - | 2 - 3 | 177 | 70 | |
AK300859.1 | - | 2 - 3 | 180 | 70 | |
AK300999.1 | - | 4 - 5 | 425 | 70 | |
BC033747.1 | - | 4 - 5 | 399 | 70 | |
BM837987.1 | - | 2 - 3 | 193 | 70 | |
DA872042.1 | - | 4 - 5 | 365 | 70 |