Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284308.1 | NP_001271237.1 | 8 - 9 | 826 | 146 |
NM_001284309.1 | NP_001271238.1 | 8 - 9 | 766 | 146 | |
NM_001284310.1 | NP_001271239.1 | 7 - 8 | 741 | 146 | |
NM_001284311.1 | NP_001271240.1 | 6 - 7 | 594 | 146 | |
NM_006869.3 | NP_006860.1 | 8 - 9 | 999 | 146 | |
GenBank mRNA | AB097049.1 | - | 8 - 9 | 973 | 146 |
AF082324.1 | - | 8 - 9 | 973 | 146 | |
AJ006422.1 | - | 8 - 9 | 995 | 146 | |
AK092471.1 | - | 7 - 8 | 743 | 146 | |
AK124979.1 | - | 6 - 7 | 845 | 146 | |
AK290517.1 | - | 9 - 10 | 945 | 146 | |
AK300644.1 | - | 6 - 7 | 521 | 146 | |
AK300859.1 | - | 6 - 7 | 581 | 146 | |
AK300999.1 | - | 8 - 9 | 826 | 146 | |
BC033747.1 | - | 8 - 9 | 800 | 146 |