Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002226.4 | NP_002217.3 | 12 - 13 | 2004 | 84 |
NM_145159.2 | NP_660142.1 | 11 - 12 | 1890 | 84 | |
GenBank mRNA | AF003521.1 | - | 12 - 13 | 1632 | 84 |
AF020201.1 | - | 12 - 13 | 1651 | 84 | |
AF029778.1 | - | 12 - 13 | 2004 | 84 | |
AF029779.1 | - | 11 - 12 | 1890 | 84 | |
BC032053.1 | - | 12 - 13 | 1668 | 84 | |
U69180.1 | - | 3 - 4 | 262 | 84 | |
Y14330.1 | - | 12 - 13 | 1555 | 84 |