Assay Design



Gene Transcripts

Gene Symbol
FAM49B
Entrez Gene ID
51571
Gene Name
family with sequence similarity 49 member B
Gene Aliases
BM-009, L1
Chromosome Location
Chr.8: 129839593 - 130016697 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256763.1 NP_001243692.1 9 - 10 1025 101
NM_016623.4 NP_057707.3 8 - 9 938 101
NR_046359.1 11 - 12 1291 101
NR_046360.1 11 - 12 1201 101
NR_046361.1 10 - 11 1191 101
XM_005250966.3 XP_005251023.1 7 - 8 794 101
XM_011517108.2 XP_011515410.1 10 - 11 1087 101
XM_011517109.2 XP_011515411.1 10 - 11 859 101
XM_011517110.2 XP_011515412.1 9 - 10 999 101
XM_011517111.2 XP_011515413.1 9 - 10 817 101
XM_011517112.2 XP_011515414.1 11 - 12 1042 101
XM_011517113.2 XP_011515415.1 10 - 11 1053 101
XM_011517114.2 XP_011515416.1 10 - 11 959 101
XM_011517115.2 XP_011515417.1 9 - 10 966 101
XM_011517116.2 XP_011515418.1 10 - 11 1000 101
XM_011517117.2 XP_011515419.1 9 - 10 969 101
XM_011517118.2 XP_011515420.1 9 - 10 913 101
XM_011517120.2 XP_011515422.1 8 - 9 912 101
XM_011517121.2 XP_011515423.1 11 - 12 924 101
XM_011517122.2 XP_011515424.1 9 - 10 1003 101
XM_011517123.2 XP_011515425.1 9 - 10 776 101
XM_011517124.2 XP_011515426.1 8 - 9 891 101
XM_011517125.2 XP_011515427.1 8 - 9 830 101
XM_017013539.1 XP_016869028.1 11 - 12 1046 101
XM_017013540.1 XP_016869029.1 11 - 12 905 101
XM_017013541.1 XP_016869030.1 9 - 10 1033 101
XM_017013542.1 XP_016869031.1 11 - 12 1083 101
XM_017013543.1 XP_016869032.1 11 - 12 1183 101
XM_017013544.1 XP_016869033.1 11 - 12 965 101
XM_017013545.1 XP_016869034.1 12 - 13 1229 101
XM_017013546.1 XP_016869035.1 10 - 11 996 101
XM_017013547.1 XP_016869036.1 12 - 13 1088 101
XM_017013548.1 XP_016869037.1 10 - 11 1092 101
XM_017013549.1 XP_016869038.1 10 - 11 955 101
XM_017013550.1 XP_016869039.1 9 - 10 1006 101
XM_017013551.1 XP_016869040.1 11 - 12 1001 101
XM_017013552.1 XP_016869041.1 11 - 12 1046 101
XM_017013553.1 XP_016869042.1 10 - 11 859 101
XM_017013554.1 XP_016869043.1 12 - 13 1071 101
XM_017013555.1 XP_016869044.1 9 - 10 1026 101
XM_017013556.1 XP_016869045.1 9 - 10 772 101
XM_017013557.1 XP_016869046.1 10 - 11 818 101
XM_017013558.1 XP_016869047.1 10 - 11 963 101
XM_017013559.1 XP_016869048.1 10 - 11 822 101
XM_017013560.1 XP_016869049.1 8 - 9 937 101
XM_017013561.1 XP_016869050.1 8 - 9 689 101
XM_017013562.1 XP_016869051.1 9 - 10 876 101
GenBank mRNA AF208851.1 - 10 - 11 877 101
AK001226.1 - 4 - 5 444 101
AK002059.1 - 11 - 12 1001 101
AK130372.1 - 8 - 9 825 101
AY364259.1 - 5 - 6 492 101
AY598320.1 - 6 - 7 624 101
BC003599.1 - 9 - 10 804 101
BC016345.1 - 10 - 11 945 101
BC017297.2 - 11 - 12 1173 101

View Full Product Details