Assay Design



Gene Transcripts

Gene Symbol
ARMC5
Entrez Gene ID
79798
Gene Name
armadillo repeat containing 5
Gene Aliases
AIMAH2
Chromosome Location
Chr.16: 31458273 - 31467167 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105247.1 NP_001098717.1 5 - 6 2528 68
NM_001288767.1 NP_001275696.1 7 - 8 2408 68
NM_001301820.1 NP_001288749.1 6 - 7 2543 68
GenBank mRNA AK294897.1 - 6 - 7 2450 68
AK295790.1 - 7 - 8 2408 68
AK297115.1 - 6 - 7 987 68
BC014945.1 - 6 - 7 1933 68

View Full Product Details