Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001194946.1 | NP_001181875.1 | 17 - 18 | 2769 | 81 |
NM_001194947.1 | NP_001181876.1 | 16 - 17 | 2712 | 81 | |
NM_001291157.1 | NP_001278086.1 | 16 - 17 | 2801 | 81 | |
NM_004953.4 | NP_004944.3 | 9 - 10 | 2260 | 81 | |
NM_182917.4 | NP_886553.3 | 15 - 16 | 2896 | 81 | |
NM_198241.2 | NP_937884.1 | 16 - 17 | 2748 | 81 | |
NM_198242.2 | NP_937885.1 | 12 - 13 | 2333 | 81 | |
NM_198244.2 | NP_937887.1 | 13 - 14 | 2510 | 81 | |
GenBank mRNA | AB210013.1 | - | 15 - 16 | 2887 | 81 |
AF104913.1 | - | 13 - 14 | 2372 | 81 | |
AF281070.1 | - | 15 - 16 | 2754 | 81 | |
AJ001046.1 | - | 3 - 4 | 437 | 81 | |
AK096719.1 | - | 14 - 15 | 2398 | 81 | |
AK128378.1 | - | 15 - 16 | 2497 | 81 | |
AK226160.1 | - | 16 - 17 | 2682 | 81 | |
AK294567.1 | - | 14 - 15 | 2379 | 81 | |
AK299763.1 | - | 13 - 14 | 2438 | 81 | |
AK308319.1 | - | 11 - 12 | 2098 | 81 | |
AL832754.1 | - | 12 - 13 | 2222 | 81 | |
AY082886.1 | - | 15 - 16 | 2754 | 81 | |
BC082764.1 | - | 9 - 10 | 1906 | 81 | |
BC140892.1 | - | 17 - 18 | 2611 | 81 | |
BC140896.1 | - | 16 - 17 | 2554 | 81 | |
BX647812.1 | - | 16 - 17 | 2661 | 81 |