Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001194946.1 | NP_001181875.1 | 18 - 19 | 2913 | 135 |
NM_001194947.1 | NP_001181876.1 | 17 - 18 | 2856 | 135 | |
NM_001291157.1 | NP_001278086.1 | 17 - 18 | 2945 | 135 | |
NM_004953.4 | NP_004944.3 | 10 - 11 | 2404 | 135 | |
NM_182917.4 | NP_886553.3 | 16 - 17 | 3040 | 135 | |
NM_198241.2 | NP_937884.1 | 17 - 18 | 2892 | 135 | |
NM_198242.2 | NP_937885.1 | 13 - 14 | 2477 | 135 | |
NM_198244.2 | NP_937887.1 | 14 - 15 | 2654 | 135 | |
GenBank mRNA | AB210013.1 | - | 16 - 17 | 3031 | 135 |
AF104913.1 | - | 14 - 15 | 2516 | 135 | |
AF281070.1 | - | 16 - 17 | 2898 | 135 | |
AJ001046.1 | - | 4 - 5 | 581 | 135 | |
AK096719.1 | - | 15 - 16 | 2542 | 135 | |
AK128378.1 | - | 16 - 17 | 2641 | 135 | |
AK226160.1 | - | 17 - 18 | 2826 | 135 | |
AK294567.1 | - | 15 - 16 | 2523 | 135 | |
AK299763.1 | - | 14 - 15 | 2582 | 135 | |
AK308319.1 | - | 12 - 13 | 2240 | 133 | |
AL832754.1 | - | 13 - 14 | 2366 | 135 | |
AY082886.1 | - | 16 - 17 | 2898 | 135 | |
BC082764.1 | - | 10 - 11 | 2050 | 135 | |
BC140892.1 | - | 18 - 19 | 2755 | 135 | |
BC140896.1 | - | 17 - 18 | 2698 | 135 | |
BX647812.1 | - | 17 - 18 | 2805 | 135 |