Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010939.2 | NP_001010939.2 | 6 - 7 | 746 | 138 |
XM_005269542.3 | XP_005269599.1 | 2 - 3 | 191 | 138 | |
XM_006717635.2 | XP_006717698.1 | 6 - 7 | 744 | 138 | |
XM_011539314.2 | XP_011537616.1 | 6 - 7 | 1269 | 138 | |
XM_011539315.2 | XP_011537617.1 | 6 - 7 | 745 | 138 | |
XM_011539317.2 | XP_011537619.1 | 6 - 7 | 1269 | 138 | |
XM_017015740.1 | XP_016871229.1 | 6 - 7 | 1269 | 138 | |
GenBank mRNA | BC031219.1 | - | 6 - 7 | 746 | 138 |