Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010939.2 | NP_001010939.2 | 9 - 10 | 1107 | 80 |
XM_005269542.3 | XP_005269599.1 | 5 - 6 | 552 | 80 | |
XM_006717635.2 | XP_006717698.1 | 9 - 10 | 1105 | 80 | |
XM_011539314.2 | XP_011537616.1 | 9 - 10 | 1630 | 80 | |
XM_011539315.2 | XP_011537617.1 | 9 - 10 | 1106 | 80 | |
XM_011539317.2 | XP_011537619.1 | 8 - 9 | 1483 | 80 | |
XM_011539318.2 | XP_011537620.1 | 4 - 5 | 757 | 80 | |
XM_011539319.1 | XP_011537621.1 | 5 - 6 | 606 | 80 | |
XM_011539320.2 | XP_011537622.1 | 5 - 6 | 602 | 80 | |
XM_017015740.1 | XP_016871229.1 | 9 - 10 | 1630 | 80 | |
GenBank mRNA | BC031219.1 | - | 9 - 10 | 1107 | 80 |