Assay Design



Gene Transcripts

Gene Symbol
BTN3A2
Entrez Gene ID
11118
Gene Name
butyrophilin subfamily 3 member A2
Gene Aliases
BT3.2, BTF4, BTN3.2, CD277
Chromosome Location
Chr.6: 26365159 - 26451697 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197246.2 NP_001184175.1 7 - 8 1014 66
NM_001197247.2 NP_001184176.1 9 - 10 1279 66
NM_001197248.2 NP_001184177.1 7 - 8 1118 66
NM_001197249.2 NP_001184178.1 8 - 9 1179 66
NM_007047.4 NP_008978.2 9 - 10 1269 66
XM_005248827.3 XP_005248884.1 9 - 10 1257 66
XM_005248831.3 XP_005248888.1 8 - 9 1056 66
XM_005248832.3 XP_005248889.1 8 - 9 1056 66
XM_006714979.3 XP_006715042.1 9 - 10 1272 66
XM_006714980.3 XP_006715043.1 9 - 10 1272 66
XM_006714981.3 XP_006715044.1 9 - 10 1262 66
XM_006714982.3 XP_006715045.1 8 - 9 1182 66
XM_011514267.2 XP_011512569.1 9 - 10 1138 66
XM_011514268.2 XP_011512570.1 9 - 10 1272 66
XM_011514269.2 XP_011512571.1 8 - 9 1041 66
XM_011514270.2 XP_011512572.1 8 - 9 975 66
XM_011514271.2 XP_011512573.1 8 - 9 841 66
XM_017010211.1 XP_016865700.1 9 - 10 1247 66
XM_017010212.1 XP_016865701.1 9 - 10 1123 66
XM_017010213.1 XP_016865702.1 8 - 9 1144 66
XM_017010214.1 XP_016865703.1 8 - 9 1167 66
XM_017010215.1 XP_016865704.1 8 - 9 975 66
XM_017010216.1 XP_016865705.1 7 - 8 966 66
XM_017010217.1 XP_016865706.1 7 - 8 885 66
GenBank mRNA AF257505.1 - 7 - 8 1016 66
AK290880.1 - 9 - 10 1207 66
AK299417.1 - 8 - 9 1063 66
AK303600.1 - 7 - 8 1035 66
BC002832.2 - 9 - 10 1160 66
BC020214.1 - 9 - 10 1160 66
U90546.1 - 9 - 10 1196 66

View Full Product Details