Assay Design



Gene Transcripts

Gene Symbol
GSN
Entrez Gene ID
2934
Gene Name
gelsolin
Gene Aliases
ADF, AGEL
Chromosome Location
Chr.9: 121201082 - 121332844 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000177.4 NP_000168.1 12 - 13 1802 83
NM_001127662.1 NP_001121134.1 12 - 13 1715 83
NM_001127663.1 NP_001121135.2 14 - 15 1920 83
NM_001127664.1 NP_001121136.1 14 - 15 1887 83
NM_001127665.1 NP_001121137.1 14 - 15 1882 83
NM_001127666.1 NP_001121138.1 14 - 15 1879 83
NM_001127667.1 NP_001121139.1 14 - 15 1870 83
NM_001258029.1 NP_001244958.1 12 - 13 1702 83
NM_001258030.1 NP_001244959.1 12 - 13 1779 83
NM_198252.2 NP_937895.1 13 - 14 1808 83
XM_005251943.1 XP_005252000.1 13 - 14 1906 83
XM_005251944.1 XP_005252001.1 13 - 14 1897 83
XM_005251945.3 XP_005252002.1 13 - 14 1810 83
XM_006717079.1 XP_006717142.1 13 - 14 1744 83
XM_011518584.1 XP_011516886.1 14 - 15 1897 83
XM_011518585.1 XP_011516887.1 12 - 13 1717 83
XM_011518586.1 XP_011516888.1 15 - 16 1920 83
XM_011518587.2 XP_011516889.1 15 - 16 1962 83
XM_011518588.2 XP_011516890.1 15 - 16 1924 83
XM_011518589.2 XP_011516891.1 15 - 16 1920 83
XM_011518590.2 XP_011516892.1 15 - 16 1872 83
XM_011518591.2 XP_011516893.1 20 - 21 2964 83
XM_011518592.1 XP_011516894.1 14 - 15 1786 83
XM_011518593.1 XP_011516895.1 15 - 16 1932 83
XM_011518594.1 XP_011516896.1 9 - 10 1129 83
XM_017014643.1 XP_016870132.1 21 - 22 3115 83
XM_017014644.1 XP_016870133.1 21 - 22 3049 83
XM_017014645.1 XP_016870134.1 13 - 14 1861 83
XM_017014646.1 XP_016870135.1 16 - 17 2032 83
XM_017014647.1 XP_016870136.1 22 - 23 3248 83
XM_017014648.1 XP_016870137.1 15 - 16 1859 83
GenBank mRNA AK092797.1 - 8 - 9 1943 83
AK096280.1 - 13 - 14 1781 83
AK125819.1 - 21 - 22 2611 83
AK295201.1 - 13 - 14 1646 83
AK295572.1 - 14 - 15 1841 83
AK297870.1 - 12 - 13 1660 83
AK299453.1 - 12 - 13 1767 83
AK300631.1 - 14 - 15 1854 83
AK304629.1 - 12 - 13 1723 83
AK304675.1 - 11 - 12 1756 83
AK307859.1 - 12 - 13 1666 83
AK315494.1 - 12 - 13 1773 83
BC017491.1 - 2 - 3 252 83
BC026033.1 - 12 - 13 1746 83
BX647999.1 - 12 - 13 1704 83
X04412.1 - 12 - 13 1755 83

View Full Product Details