Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011649.2 | NP_001011649.1 | 12 - 13 | 1505 | 96 |
NM_001272039.1 | NP_001258968.1 | 12 - 13 | 1505 | 96 | |
NM_018249.5 | NP_060719.4 | 12 - 13 | 1505 | 96 | |
NR_073554.1 | 12 - 13 | 1505 | 96 | ||
NR_073555.1 | 12 - 13 | 1505 | 96 | ||
NR_073557.1 | 12 - 13 | 1505 | 96 | ||
NR_073558.1 | 12 - 13 | 1502 | 96 | ||
XM_006717182.1 | XP_006717245.1 | 12 - 13 | 1494 | 96 | |
XM_006717185.1 | XP_006717248.1 | 12 - 13 | 1494 | 96 | |
XM_011518860.1 | XP_011517162.1 | 12 - 13 | 1494 | 96 | |
XM_017014921.1 | XP_016870410.1 | 12 - 13 | 1494 | 96 | |
XM_017014922.1 | XP_016870411.1 | 6 - 7 | 723 | 96 | |
XM_017014923.1 | XP_016870412.1 | 12 - 13 | 1494 | 96 | |
XM_017014924.1 | XP_016870413.1 | 3 - 4 | 399 | 96 | |
GenBank mRNA | AB046853.2 | - | 12 - 13 | 1474 | 96 |
AF448860.1 | - | 12 - 13 | 1455 | 96 | |
AK122913.1 | - | 12 - 13 | 1494 | 96 | |
AL833481.1 | - | 12 - 13 | 1493 | 96 | |
BC136275.1 | - | 12 - 13 | 1340 | 96 | |
BC143762.1 | - | 12 - 13 | 1340 | 96 | |
BC146782.1 | - | 12 - 13 | 1474 | 96 | |
BX537421.1 | - | 12 - 13 | 1490 | 96 | |
BX537708.1 | - | 12 - 13 | 1494 | 96 | |
BX537759.1 | - | 12 - 13 | 1493 | 96 | |
BX640896.1 | - | 6 - 7 | 717 | 96 | |
CR936622.1 | - | 5 - 6 | 635 | 96 | |
CR936717.1 | - | 5 - 6 | 635 | 96 | |
KP294527.1 | - | 12 - 13 | 1505 | 96 |