Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172810.1 | NP_001166281.1 | 3 - 4 | 452 | 108 |
NM_023036.4 | NP_075462.3 | 3 - 4 | 455 | 108 | |
XM_011525125.2 | XP_011523427.1 | 3 - 4 | 472 | 108 | |
GenBank mRNA | AF250288.1 | - | 3 - 4 | 412 | 108 |
AJ295276.1 | - | 2 - 3 | 651 | 108 | |
BC039582.1 | - | 3 - 4 | 442 | 108 | |
BI462861.1 | - | 3 - 4 | 451 | 108 | |
BI464166.1 | - | 3 - 4 | 441 | 108 |