Assay Design



Gene Transcripts

Gene Symbol
DNAI2
Entrez Gene ID
64446
Gene Name
dynein axonemal intermediate chain 2
Gene Aliases
CILD9, DIC2
Chromosome Location
Chr.17: 74274215 - 74314884 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172810.1 NP_001166281.1 8 - 9 1096 56
NM_023036.4 NP_075462.3 8 - 9 1099 56
XM_011525125.2 XP_011523427.1 8 - 9 1116 56
GenBank mRNA AF250288.1 - 8 - 9 1056 56
AJ295276.1 - 7 - 8 1295 56
BC033013.1 - 7 - 8 973 56
BC039582.1 - 8 - 9 1086 56

View Full Product Details