Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001123355.1 | NP_001116827.1 | 5 - 6 | 709 | 94 |
NM_001123369.1 | NP_001116841.1 | 3 - 4 | 532 | 94 | |
NM_002721.4 | NP_002712.1 | 4 - 5 | 598 | 94 | |
XM_011518847.2 | XP_011517149.1 | 4 - 5 | 395 | 94 | |
GenBank mRNA | AF035158.2 | - | 4 - 5 | 371 | 94 |
AK295190.1 | - | 5 - 6 | 574 | 94 | |
AK299087.1 | - | 3 - 4 | 396 | 94 | |
AK312332.1 | - | 4 - 5 | 430 | 94 | |
BC006990.1 | - | 4 - 5 | 445 | 94 | |
BT019708.1 | - | 4 - 5 | 377 | 94 | |
DC334216.1 | - | 5 - 6 | 572 | 94 | |
DC369815.1 | - | 3 - 4 | 396 | 94 | |
KU178271.1 | - | 4 - 5 | 377 | 94 | |
X92972.2 | - | 4 - 5 | 398 | 94 |