Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000834.3 | NP_000825.2 | 10 - 11 | 2346 | 121 |
XM_011520628.2 | XP_011518930.1 | 11 - 12 | 3139 | 121 | |
XM_011520629.2 | XP_011518931.1 | 12 - 13 | 2946 | 121 | |
XM_017019219.1 | XP_016874708.1 | 11 - 12 | 2694 | 121 | |
GenBank mRNA | AB208850.1 | - | 7 - 8 | 1125 | 121 |
BC113618.1 | - | 10 - 11 | 2245 | 121 | |
BC113620.1 | - | 10 - 11 | 2245 | 121 | |
U11287.1 | - | 10 - 11 | 2377 | 121 | |
U28862.1 | - | 2 - 3 | 187 | 121 | |
U88963.1 | - | 10 - 11 | 2346 | 121 | |
U90278.1 | - | 10 - 11 | 2376 | 121 |