Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000834.3 | NP_000825.2 | 2 - 3 | 589 | 110 |
XM_011520628.2 | XP_011518930.1 | 3 - 4 | 1382 | 110 | |
XM_011520629.2 | XP_011518931.1 | 4 - 5 | 1189 | 110 | |
XM_017019219.1 | XP_016874708.1 | 3 - 4 | 937 | 110 | |
GenBank mRNA | BC113618.1 | - | 2 - 3 | 488 | 110 |
BC113620.1 | - | 2 - 3 | 488 | 110 | |
U11287.1 | - | 2 - 3 | 620 | 110 | |
U28861.1 | - | 1 - 2 | 410 | 110 | |
U88963.1 | - | 2 - 3 | 589 | 110 | |
U90278.1 | - | 2 - 3 | 619 | 110 |