Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004019.1 | NP_001004019.1 | 13 - 14 | 79 | |
NM_001165035.1 | NP_001158507.1 | 13 - 14 | 79 | ||
NM_001998.2 | NP_001989.2 | 12 - 13 | 79 | ||
XM_006713026.2 | XP_006713089.1 | 12 - 13 | 79 | ||
GenBank mRNA | AK129883.1 | - | - | 2307 | 79 |
AK300913.1 | - | 8 - 9 | 973 | 79 | |
AK304777.1 | - | 12 - 13 | 79 | ||
AK304827.1 | - | 13 - 14 | 79 | ||
AL050095.1 | - | 7 - 8 | 79 | ||
AY130459.1 | - | 13 - 14 | 2907 | 79 | |
BC051690.1 | - | 12 - 13 | 2785 | 79 | |
X82494.1 | - | 12 - 13 | 2766 | 79 |