Assay Design



Gene Transcripts

Gene Symbol
FBLN2
Entrez Gene ID
2199
Gene Name
fibulin 2
Gene Aliases
-
Chromosome Location
Chr.3: 13549125 - 13638422 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004019.1 NP_001004019.1 15 - 16 3210 73
NM_001165035.1 NP_001158507.1 15 - 16 3467 73
NM_001998.2 NP_001989.2 14 - 15 3069 73
GenBank mRNA AK129883.1 - - 2554 73
AK300913.1 - 10 - 11 1220 73
AK304777.1 - 14 - 15 3169 73
AK304827.1 - 15 - 16 3467 73
AL050095.1 - 9 - 10 1121 73
AY130459.1 - 15 - 16 3154 73
BC051690.1 - 14 - 15 3032 73
X82494.1 - 14 - 15 3013 73

View Full Product Details