Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004019.1 | NP_001004019.1 | 5 - 6 | 1856 | 94 |
NM_001165035.1 | NP_001158507.1 | 5 - 6 | 2113 | 94 | |
NM_001998.2 | NP_001989.2 | 5 - 6 | 1856 | 94 | |
XM_006713026.2 | XP_006713089.1 | 4 - 5 | 1765 | 94 | |
GenBank mRNA | AK129883.1 | - | - | 295 | 94 |
AK304777.1 | - | 5 - 6 | 1956 | 94 | |
AK304827.1 | - | 5 - 6 | 2113 | 94 | |
AY130459.1 | - | 5 - 6 | 1800 | 94 | |
BC051690.1 | - | 5 - 6 | 1819 | 94 | |
X82494.1 | - | 5 - 6 | 1800 | 94 |